Assay Details
| Gene Symbol: SDH1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126074.3 | NP_201477.1 | 5 - 6 | 822 | 96 | |
| GenBank mRNA | AF367341.1 | - | 5 - 6 | 725 | 96 | |
| AF436833.1 | - | 5 - 6 | 771 | 96 | ||
| AY045674.1 | - | 5 - 6 | 723 | 96 | ||
| AY124812.1 | - | 5 - 6 | 653 | 96 | ||
More Information
Set Membership: |
|

SDH1-1
Genomic Map