Assay Details
| Gene Symbol: TOE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345814.1 | NP_001332132.1 | 3 - 4 | 731 | 125 | |
| NM_001345815.1 | NP_001332130.1 | 3 - 4 | 709 | 125 | ||
| NM_001345816.1 | NP_001332131.1 | 3 - 4 | 709 | 125 | ||
| NM_126118.4 | NP_201519.1 | 3 - 4 | 709 | 125 | ||
| GenBank mRNA | AK176145.1 | - | 3 - 4 | 709 | 125 | |
| AY088238.1 | - | 3 - 4 | 480 | 125 | ||
| BT029206.1 | - | 3 - 4 | 394 | 125 | ||
More Information
Set Membership: |
|

TOE3
Genomic Map