Assay Details
| Gene Symbol: TOE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345814.1 | NP_001332132.1 | 4 - 5 | 824 | 131 | |
| NM_001345815.1 | NP_001332130.1 | 4 - 5 | 802 | 131 | ||
| NM_001345816.1 | NP_001332131.1 | 4 - 5 | 802 | 131 | ||
| NM_126118.4 | NP_201519.1 | 4 - 5 | 802 | 131 | ||
| GenBank mRNA | AK176145.1 | - | 4 - 5 | 802 | 131 | |
| AY088238.1 | - | 4 - 5 | 573 | 131 | ||
| BT029206.1 | - | 4 - 5 | 487 | 131 | ||
More Information
Set Membership: |
|

TOE3
Genomic Map