Assay Details
| Gene Symbol: TOE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345814.1 | NP_001332132.1 | 7 - 8 | 1110 | 110 | |
| NM_126118.4 | NP_201519.1 | 7 - 8 | 1088 | 110 | ||
| GenBank mRNA | AK176145.1 | - | 7 - 8 | 1088 | 110 | |
| AY088238.1 | - | 7 - 8 | 859 | 110 | ||
| BT029206.1 | - | 7 - 8 | 773 | 110 | ||
More Information
Set Membership: |
|

TOE3
Genomic Map