Assay Details
| Gene Symbol: TOE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126118.4 | NP_201519.1 | 8 - 9 | 1160 | 146 | |
| GenBank mRNA | AK176145.1 | - | 8 - 9 | 1160 | 146 | |
| AY088238.1 | - | 8 - 9 | 931 | 146 | ||
| BT029206.1 | - | 8 - 9 | 845 | 146 | ||
More Information
Set Membership: |
|

TOE3
Genomic Map