Assay Details
| Gene Symbol: DEAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126119.2 | NP_201520.1 | 1 | 911 | 100 | |
| GenBank mRNA | BT025744.1 | - | 1 | 484 | 100 | |
| DQ447114.1 | - | 1 | 484 | 100 | ||
More Information
Set Membership: |
|

DEAR2
Genomic Map