Assay Details
| Gene Symbol: SKIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126125.3 | NP_569047.1 | 1 | 2195 | 112 | |
| GenBank mRNA | AF263378.1 | - | 1 | 1603 | 112 | |
| BT002473.1 | - | 1 | 1625 | 112 | ||
More Information
Set Membership: |
|

SKIP2
Genomic Map