Assay Details
| Gene Symbol: EB1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126127.4 | NP_201528.1 | 3 - 4 | 493 | 96 | |
| GenBank mRNA | AK175495.1 | - | 3 - 4 | 437 | 96 | |
| AY087775.1 | - | 3 - 4 | 470 | 96 | ||
| BT028930.1 | - | 3 - 4 | 373 | 96 | ||
More Information
Set Membership: |
|

EB1C
Genomic Map