Assay Details
| Gene Symbol: NPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126043.1 | NP_001119515.1 | 2 - 3 | 667 | 227 | |
| NM_001345830.1 | NP_001331087.1 | 2 - 3 | 444 | 149 | ||
| NM_126144.4 | NP_201545.1 | 2 - 3 | 826 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001126043.1 transcripts.
Set Membership: |
|

NPY3
Genomic Map