Assay Details
| Gene Symbol: NPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345830.1 | NP_001331087.1 | 3 - 4 | 520 | 115 | |
| NM_126144.4 | NP_201545.1 | 3 - 4 | 902 | 115 | ||
More Information
Set Membership: |
|

NPY3
Genomic Map