Assay Details
| Gene Symbol: PDE345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126176.5 | NP_178224.1 | 1 - 2 | 528 | 75 | |
| GenBank mRNA | AF325014.2 | - | 1 - 2 | 254 | 75 | |
| AY086203.1 | - | 1 - 2 | 254 | 75 | ||
More Information
Set Membership: |
|

PDE345
Genomic Map