Assay Details
| Gene Symbol: BAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126178.4 | NP_565254.1 | 2 - 3 | 476 | 61 | |
| NM_201658.1 | NP_973387.1 | 1 - 2 | 485 | 61 | ||
| GenBank mRNA | AF019637.1 | - | 2 - 3 | 395 | 61 | |
| BT020534.1 | - | 1 - 2 | 163 | 61 | ||
More Information
Set Membership: |
|

BAT1
Genomic Map