Assay Details
Gene Symbol: HB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335062.1 | NP_001324758.1 | 1 - 2 | 295 | 59 | |
NM_126204.3 | NP_178252.2 | 1 - 2 | 295 | 59 | ||
GenBank mRNA | AJ431181.1 | - | 1 - 2 | 295 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
