Assay Details
| Gene Symbol: PAHX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335065.1 | NP_001325094.1 | 2 - 3 | 480 | 77 | |
| NM_126210.4 | NP_565262.1 | 2 - 3 | 487 | 77 | ||
| GenBank mRNA | AY057710.1 | - | 2 - 3 | 204 | 77 | |
| AY116965.1 | - | 2 - 3 | 184 | 77 | ||
More Information
Set Membership: |
|

PAHX
Genomic Map