Assay Details
| Gene Symbol: PAHX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335065.1 | NP_001325094.1 | 3 - 4 | 540 | 138 | |
| NM_126210.4 | NP_565262.1 | 3 - 4 | 547 | 138 | ||
| GenBank mRNA | AY057710.1 | - | 3 - 4 | 264 | 138 | |
| AY116965.1 | - | 3 - 4 | 244 | 138 | ||
More Information
Set Membership: |
|

PAHX
Genomic Map