Assay Details
| Gene Symbol: PAHX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335065.1 | NP_001325094.1 | 4 - 5 | 600 | 120 | |
| NM_126210.4 | NP_565262.1 | 4 - 5 | 607 | 120 | ||
| GenBank mRNA | AY057710.1 | - | 4 - 5 | 324 | 120 | |
| AY116965.1 | - | 4 - 5 | 304 | 120 | ||
More Information
Set Membership: |
|

PAHX
Genomic Map