Assay Details
| Gene Symbol: WOL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335080.1 | NP_001324428.1 | 1 - 2 | 1466 | 80 | |
| NM_001335081.1 | NP_001324427.1 | 2 - 3 | 943 | 80 | ||
| NM_001335082.1 | NP_001324429.1 | 2 - 3 | 652 | 80 | ||
| NM_126244.3 | NP_565277.1 | 2 - 3 | 969 | 80 | ||
| NM_179594.2 | NP_849925.1 | 2 - 3 | 948 | 80 | ||
| NM_201667.1 | NP_973396.1 | 3 - 4 | 669 | 80 | ||
| GenBank mRNA | AB046871.1 | - | 2 - 3 | 607 | 80 | |
| AJ278528.1 | - | 2 - 3 | 677 | 80 | ||
| AJ278529.1 | - | 2 - 3 | 560 | 80 | ||
| AJ278530.1 | - | 3 - 4 | 669 | 80 | ||
More Information
Set Membership: |
|

WOL
Genomic Map