Assay Details
| Gene Symbol: WOL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335080.1 | NP_001324428.1 | 7 - 8 | 2669 | 140 | |
| NM_001335081.1 | NP_001324427.1 | 8 - 9 | 2146 | 140 | ||
| NM_001335082.1 | NP_001324429.1 | 8 - 9 | 1855 | 140 | ||
| NM_126244.3 | NP_565277.1 | 8 - 9 | 2172 | 140 | ||
| NM_179594.2 | NP_849925.1 | 8 - 9 | 2151 | 140 | ||
| NM_201667.1 | NP_973396.1 | 9 - 10 | 1872 | 140 | ||
| GenBank mRNA | AB046871.1 | - | 8 - 9 | 1810 | 140 | |
| AJ278528.1 | - | 8 - 9 | 1880 | 140 | ||
| AJ278529.1 | - | 8 - 9 | 1763 | 140 | ||
| AJ278530.1 | - | 9 - 10 | 1872 | 140 | ||
More Information
Set Membership: |
|

WOL
Genomic Map