Assay Details
| Gene Symbol: ETL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126270.2 | NP_178318.1 | 5 - 6 | 1429 | 68 | |
| GenBank mRNA | AY059857.1 | - | 5 - 6 | 1368 | 68 | |
| BT002576.1 | - | 5 - 6 | 1171 | 68 | ||
More Information
Set Membership: |
|

ETL1
Genomic Map