Assay Details
| Gene Symbol: ETL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126270.2 | NP_178318.1 | 8 - 9 | 1952 | 87 | |
| GenBank mRNA | AY059857.1 | - | 8 - 9 | 1891 | 87 | |
| BT002576.1 | - | 8 - 9 | 1694 | 87 | ||
More Information
Set Membership: |
|

ETL1
Genomic Map