Assay Details
| Gene Symbol: TOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126278.3 | NP_027422.1 | 2 - 3 | 574 | 80 | |
| GenBank mRNA | AB036427.1 | - | 2 - 3 | 312 | 80 | |
| AK117222.1 | - | 2 - 3 | 341 | 80 | ||
More Information
Set Membership: |
|

TOM3
Genomic Map