Assay Details
| Gene Symbol: PLPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335131.1 | NP_001318183.1 | 2 - 3 | 1202 | 148 | |
| NM_001335132.1 | NP_001324185.1 | 2 - 3 | 1202 | 148 | ||
| NM_126326.5 | NP_565288.1 | 2 - 3 | 1242 | 148 | ||
| NM_179597.1 | NP_849928.1 | 2 - 3 | 1191 | 148 | ||
| GenBank mRNA | AB038798.1 | - | 2 - 3 | 1192 | 148 | |
| AY045642.1 | - | 2 - 3 | 994 | 148 | ||
| AY058232.1 | - | 2 - 3 | 786 | 148 | ||
More Information
Set Membership: |
|

PLPB
Genomic Map