Assay Details
| Gene Symbol: APK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126335.6 | NP_178383.1 | 4 - 5 | 1019 | 133 | |
| NM_201674.2 | NP_973403.1 | 4 - 5 | 870 | 133 | ||
| GenBank mRNA | AF428432.1 | - | 4 - 5 | 869 | 133 | |
| AY125559.1 | - | 4 - 5 | 678 | 133 | ||
More Information
Set Membership: |
|

APK2B
Genomic Map