Assay Details
| Gene Symbol: MYB88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035880.3 | NP_001030957.1 | 1 - 2 | 857 | 85 | |
| NM_001335147.1 | NP_001325364.1 | 1 - 2 | 718 | 85 | ||
| NM_126337.4 | NP_565291.2 | 1 - 2 | 857 | 85 | ||
More Information
Set Membership: |
|

MYB88
Genomic Map