Assay Details
| Gene Symbol: PKS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126347.3 | NP_565292.1 | 1 | 1442 | 105 | |
| GenBank mRNA | AF325064.1 | - | 1 | 1185 | 105 | |
| AY052708.1 | - | 1 | 1230 | 105 | ||
| AY063721.1 | - | 1 | 1185 | 105 | ||
More Information
Set Membership: |
|

PKS1
Genomic Map