Assay Details
| Gene Symbol: AT2G02970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126349.3 | NP_565293.1 | 3 - 4 | 1129 | 94 | |
| GenBank mRNA | JF830011.1 | - | 3 - 4 | 844 | 94 | |
More Information
Set Membership: |
|

AT2G02970
Genomic Map