Assay Details
| Gene Symbol: AT2G02970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126349.3 | NP_565293.1 | 4 - 5 | 1187 | 125 | |
| GenBank mRNA | JF830011.1 | - | 4 - 5 | 902 | 125 | |
More Information
Set Membership: |
|

AT2G02970
Genomic Map