Assay Details
| Gene Symbol: emb1579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335170.1 | NP_001323719.1 | 9 - 10 | 1902 | 101 | |
| NM_126366.5 | NP_178414.2 | 9 - 10 | 1944 | 101 | ||
More Information
Set Membership: |
|

emb1579
Genomic Map