Assay Details
| Gene Symbol: SK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126368.2 | NP_565296.1 | 1 | 241 | 65 | |
| GenBank mRNA | DQ056526.1 | - | 1 | 227 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SK14| Gene Symbol: SK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126368.2 | NP_565296.1 | 1 | 241 | 65 | |
| GenBank mRNA | DQ056526.1 | - | 1 | 227 | 65 | |
Set Membership: |
|