Assay Details
| Gene Symbol: SK16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126370.2 | NP_565297.1 | 1 | 261 | 120 | |
| GenBank mRNA | BT015070.1 | - | 1 | 257 | 120 | |
| BT015726.1 | - | 1 | 243 | 120 | ||
More Information
Set Membership: |
|

SK16
Genomic Map