Assay Details
| Gene Symbol: CFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111003.5 | NP_186786.2 | 12 - 13 | 2840 | 72 | |
| GenBank mRNA | AY136347.1 | - | 12 - 13 | 2840 | 72 | |
| BT010594.1 | - | 12 - 13 | 2818 | 72 | ||
More Information
Set Membership: |
|

CFM2
Genomic Map