Assay Details
| Gene Symbol: CFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111003.5 | NP_186786.2 | 1 - 2 | 710 | 97 | |
| GenBank mRNA | AY136347.1 | - | 1 - 2 | 710 | 97 | |
| BT010594.1 | - | 1 - 2 | 688 | 97 | ||
More Information
Set Membership: |
|

CFM2
Genomic Map