Assay Details
| Gene Symbol: DOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111008.3 | NP_186791.1 | 8 - 9 | 1636 | 119 | |
| GenBank mRNA | AY042787.1 | - | 8 - 9 | 1568 | 119 | |
| AY064666.1 | - | 8 - 9 | 1457 | 119 | ||
More Information
Set Membership: |
|

DOX1
Genomic Map