Assay Details
| Gene Symbol: CDC48C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337349.1 | NP_001326134.1 | 1 - 2 | 1060 | 89 | |
| NM_111027.4 | NP_186810.2 | 1 - 2 | 1101 | 89 | ||
| GenBank mRNA | AX765816.1 | - | 1 - 2 | 924 | 89 | |
| AY136443.1 | - | 1 - 2 | 957 | 89 | ||
More Information
Set Membership: |
|

CDC48C
Genomic Map