Assay Details
| Gene Symbol: CDC48C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337349.1 | NP_001326134.1 | 2 - 3 | 1649 | 110 | |
| NM_111027.4 | NP_186810.2 | 2 - 3 | 1690 | 110 | ||
| GenBank mRNA | AX765816.1 | - | 2 - 3 | 1513 | 110 | |
| AY136443.1 | - | 2 - 3 | 1546 | 110 | ||
More Information
Set Membership: |
|

CDC48C
Genomic Map