Assay Details
| Gene Symbol: CDC48C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337349.1 | NP_001326134.1 | 3 - 4 | 1803 | 122 | |
| NM_111027.4 | NP_186810.2 | 3 - 4 | 1844 | 122 | ||
| GenBank mRNA | AX765816.1 | - | 3 - 4 | 1667 | 122 | |
| AY136443.1 | - | 3 - 4 | 1700 | 122 | ||
More Information
Set Membership: |
|

CDC48C
Genomic Map