Assay Details
| Gene Symbol: SRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111070.4 | NP_566159.1 | 1 - 2 | 345 | 73 | |
| GenBank mRNA | AY072127.1 | - | 1 - 2 | 313 | 73 | |
| AY114073.1 | - | 1 - 2 | 244 | 73 | ||
More Information
Set Membership: |
|

SRG3
Genomic Map