Assay Details
| Gene Symbol: BIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337388.1 | NP_001327521.1 | 10 - 11 | 12944 | 66 | |
| NM_001337389.1 | NP_001327522.1 | 10 - 11 | 12872 | 66 | ||
| NM_001337390.1 | NP_001327520.1 | 10 - 11 | 12815 | 66 | ||
| NM_111093.2 | NP_186875.2 | 10 - 11 | 12816 | 66 | ||
More Information
Set Membership: |
|

BIG
Genomic Map