Assay Details
| Gene Symbol: BIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337390.1 | NP_001327520.1 | 1 - 2 | 353 | 101 | |
| NM_111093.2 | NP_186875.2 | 1 - 2 | 321 | 101 | ||
More Information
Set Membership: |
|

BIG
Genomic Map