Assay Details
| Gene Symbol: BIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111093.2 | NP_186875.2 | 7 - 8 | 7675 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BIG| Gene Symbol: BIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111093.2 | NP_186875.2 | 7 - 8 | 7675 | 78 | |
Set Membership: |
|