Assay Details
| Gene Symbol: AT3G02530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111120.3 | NP_186902.1 | 13 - 14 | 1566 | 74 | |
| GenBank mRNA | AY062729.1 | - | 13 - 14 | 1536 | 74 | |
| AY114652.1 | - | 13 - 14 | 1411 | 74 | ||
| BT001156.1 | - | 13 - 14 | 1411 | 74 | ||
More Information
Set Membership: |
|

AT3G02530
Genomic Map