Assay Details
| Gene Symbol: AT3G02710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111139.2 | NP_186920.2 | 2 - 3 | 1236 | 111 | |
| GenBank mRNA | BT030320.1 | - | 2 - 3 | 1165 | 111 | |
More Information
Set Membership: |
|

AT3G02710
Genomic Map