Assay Details
| Gene Symbol: AT3G02710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111139.2 | NP_186920.2 | 3 - 4 | 1327 | 70 | |
| GenBank mRNA | BT030320.1 | - | 3 - 4 | 1256 | 70 | |
More Information
Set Membership: |
|

AT3G02710
Genomic Map