Assay Details
| Gene Symbol: TRXF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111141.3 | NP_186922.1 | 2 - 3 | 491 | 128 | |
| GenBank mRNA | AY065391.1 | - | 2 - 3 | 473 | 128 | |
| AY084778.1 | - | 2 - 3 | 483 | 128 | ||
| AY096721.1 | - | 2 - 3 | 389 | 128 | ||
More Information
Set Membership: |
|

TRXF1
Genomic Map