Assay Details
| Gene Symbol: IPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337434.1 | NP_001325698.1 | 4 - 5 | 755 | 74 | |
| NM_111146.4 | NP_186927.1 | 4 - 5 | 1318 | 74 | ||
| GenBank mRNA | AF428343.1 | - | 4 - 5 | 487 | 74 | |
| AY127023.1 | - | 4 - 5 | 479 | 74 | ||
| AY143845.1 | - | 4 - 5 | 460 | 74 | ||
| U49259.1 | - | 4 - 5 | 477 | 74 | ||
More Information
Set Membership: |
|

IPP2
Genomic Map