Assay Details
| Gene Symbol: IPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337434.1 | NP_001325698.1 | 5 - 6 | 896 | 63 | |
| NM_111146.4 | NP_186927.1 | 5 - 6 | 1459 | 63 | ||
| GenBank mRNA | AF428343.1 | - | 5 - 6 | 628 | 63 | |
| AY127023.1 | - | 5 - 6 | 620 | 63 | ||
| AY143845.1 | - | 5 - 6 | 601 | 63 | ||
| U49259.1 | - | 5 - 6 | 618 | 63 | ||
More Information
Set Membership: |
|

IPP2
Genomic Map