Assay Details
| Gene Symbol: PFA-DSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111148.3 | NP_186929.2 | 2 - 3 | 449 | 147 | |
| GenBank mRNA | AK175475.1 | - | 2 - 3 | 427 | 147 | |
More Information
Set Membership: |
|

PFA-DSP3
Genomic Map