Assay Details
| Gene Symbol: ZFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337436.1 | NP_001325699.1 | 3 - 4 | 790 | 103 | |
| NM_111151.5 | NP_566183.1 | 4 - 5 | 693 | 103 | ||
| GenBank mRNA | AK117978.1 | - | 4 - 5 | 594 | 103 | |
| BT025966.1 | - | 4 - 5 | 390 | 103 | ||
More Information
Set Membership: |
|

ZFN1
Genomic Map