Assay Details
| Gene Symbol: AT3G02840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111152.2 | NP_566184.1 | 1 | 763 | 127 | |
| GenBank mRNA | AK118550.1 | - | 1 | 763 | 127 | |
| BT002533.1 | - | 1 | 763 | 127 | ||
| BT008474.1 | - | 1 | 692 | 127 | ||
More Information
Set Membership: |
|

AT3G02840
Genomic Map