Assay Details
| Gene Symbol: AT3G03140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111184.5 | NP_186964.1 | 2 - 3 | 1202 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G03140| Gene Symbol: AT3G03140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111184.5 | NP_186964.1 | 2 - 3 | 1202 | 132 | |
Set Membership: |
|